Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Gold standard solution, for AAS, 1mg/mL Au in 0.5N HCl, ACROS Organics™
Click to view available options
Quantity:
100mL
500mL
Packaging:
Plastic bottle
Description
For atomic adsorption spectroscopy
Specifications
Solubility | Solubility in water: soluble. |
Concentration | 990 to 1010μg/mL of Au(3+) in 10 to 20% HCl |
Formula Weight | 196.97 |
Physical Form | Powder and/or Chunks |
Grade | AAS |
Packaging | Plastic bottle |
Density | 1.0900g/mL |
Color | Orange-Yellow to Yellow |
Quantity | 100mL |
Specific Gravity | 1.09 |
Show More |
Safety and Handling
GHS H Statement
May be corrosive to metals. Causes severe skin burns and eye damage. May cause respiratory irritation.
GHS P Statement
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing. Immediately call a POISON CENTER or doctor/physician. Wear protective gloves/protective clothing/eye protection/face protection. IF SWALLOWED: Rinse mouth. Do NOT induce vomiting. IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/shower. IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Danger