Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Gold standard solution, for AAS, 1mg/mL Au in 0.5N HCl, ACROS Organics™

Click to view available options
Quantity:
100mL
500mL
Packaging:
Plastic bottle
7440-57-5,7647-01-0,7732-18-5
Au
196.967
MFCD00003436
PCHJSUWPFVWCPO-UHFFFAOYSA-N
23985
CHEBI:30050
gold
[Au]
7440-57-5,7647-01-0,7732-18-5
Au
196.967
MFCD00003436
PCHJSUWPFVWCPO-UHFFFAOYSA-N
23985
CHEBI:30050
gold
[Au]
For atomic adsorption spectroscopy
Solubility Solubility in water: soluble.
Concentration 990 to 1010μg/mL of Au(3+) in 10 to 20% HCl
Formula Weight 196.97
Physical Form Powder and/or Chunks
Grade AAS
Packaging Plastic bottle
Density 1.0900g/mL
Color Orange-Yellow to Yellow
Quantity 500mL
Specific Gravity 1.09
Chemical Name or Material Gold standard solution, 1 mg/ml Au in 0.5N HCl
Show More Show Less

GHS H Statement
May be corrosive to metals. Causes severe skin burns and eye damage. May cause respiratory irritation.

GHS P Statement
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing. Immediately call a POISON CENTER or doctor/physician. Wear protective gloves/protective clothing/eye protection/face protection. IF SWALLOWED: Rinse mouth. Do NOT induce vomiting. IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/shower. IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.

Danger