Alfa Aesar™ Gold Ultrathin foil, 1.0 micron thick, 99.9% (metals basis)

Manufacturer: Alfa Aesar™ 40738-FF

Quantity 25x25mm

 View more versions of this product

Catalog No. AA40738FF

 N/A
N/A

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000. 


    Chemical Identifiers

    Chemical Name or Material Gold Ultrathin foil, Temporary acrylic support

    Specifications

    Physical Form Foil
    Odor Odorless
    Particle Size 1.0 Micron thick
    TSCA TSCA
    Quantity 25x25mm
    Chemical Name or Material Gold Ultrathin foil, Temporary acrylic support
    Assay 99.9%, (metals basis)

  • Description & Specifications

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit