Alfa Aesar™ Gold wire, 0.5mm (0.02 in.) dia., annealed, Premion™, 99.999% (metals basis)

Manufacturer: Alfa Aesar™ 47160CB

Quantity 250cm

 View more versions of this product

Catalog No. AA47160CB

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 7440-57-5
    MDL Number MFCD00003436
    Chemical Name or Material Gold wire, 0.5mm (0.02 in.) dia., annealed
    Grade Premion™

    Specifications

    CAS 7440-57-5
    TSCA TSCA
    MDL Number MFCD00003436
    Odor Odorless
    Color Gold
    Weight ≈3.79g/m
    Quantity 250cm
    RTECS Number MD5070000
    Chemical Name or Material Gold wire, 0.5mm (0.02 in.) dia., annealed
    Assay 99.999%, (metals basis)
    Grade Premion™

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit