Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ Gold wire, 14kt, yellow, 1.63mm (0.064 in.) dia., Au 58.3% min

Manufacturer:  Alfa Aesar™ 40329BU

Quantity 25cm

 View more versions of this product

Catalog No. AA40329BU


  Check Availability
Add to cart

Specifications Description & Specifications

Chemical Identifiers

MFCD00003436
Gold wire (14kt), 1.63mm (0.064 in.) dia.

Specifications

≈28g/m
% Concentration Cu>Zn>Ag, Au ≤58.3%
MFCD00003436
Gold wire (14kt), 1.63mm (0.064 in.) dia.
25cm
Wire
Yellow

Safety and Handling

SDS & Safety Documents

TSCA : TSCA

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit