Alfa Aesar™ Gold wire, 14kt, yellow, 1.63mm (0.064 in.) dia., Au 58.3% min

Manufacturer: Alfa Aesar™ 40329BW

Quantity 50cm

 View more versions of this product

Catalog No. AA40329BW

  Check Availability
Add to Cart

    Chemical Identifiers

    MDL Number MFCD00003436
    Chemical Name or Material Gold wire (14kt), 1.63mm (0.064 in.) dia.

    Specifications

    Concentration % Concentration Cu>Zn>Ag, Au ≤58.3%
    Physical Form Wire
    Color Yellow
    Weight ≈28g/m
    Quantity 50cm
    TSCA TSCA
    MDL Number MFCD00003436
    Chemical Name or Material Gold wire (14kt), 1.63mm (0.064 in.) dia.

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit