Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Gold wire, 14kt, yellow, 1.63mm (0.064 in.) dia., Au 58.3% min, Thermo Scientific Chemicals

$528.00 - $1051.00

Specifications

Chemical Name or Material Gold wire (14kt)
Color Yellow
Name Note 1.63mm (0.064 in.) dia.
MDL Number MFCD00003436
Solubility Information Soluble in aqua regia,nitric acid and hydrochloric acid. Insoluble in water.
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
AA40329BS
View Documents
Thermo Scientific Chemicals
040329BS
10 cm
Each for $528.00
Order in Bulk

Request bulk or custom formats

Add to Cart
 
AA40329BU
View Documents
Thermo Scientific Chemicals
040329BU
25 cm
Each for $1,051.00
Order in Bulk

Request bulk or custom formats

Add to Cart
 
Description

Description

Gold wire is used in jewelry and as a plated coating on a wide variety of electrical and mechanical components. It is also acts as a heat reflecting coating for glass as well as a decorative medium.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

Gold wire (14kt)
1.63mm (0.064 in.) dia.
Soluble in aqua regia,nitric acid and hydrochloric acid. Insoluble in water.
≈28g/m
Yellow
MFCD00003436
% Concentration Cu>Zn>Ag, Au ≤58.3%
Wire
Safety and Handling

Safety and Handling

TSCA : Yes

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Product Certifications

RUO – Research Use Only

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit