Sartorius™ GoldLINE Electrodes

Superior stable results due to patented three chamber Iodine/Iodide metal-ion free reference system that lower temperature sensitivities and provides a metal ion free reference system ideal for Pharma and R&D grade application.

Manufacturer: Sartorius™ PYP292S

 View more versions of this product

Catalog No. 14-558-080

  Check Availability
Add to Cart

Includes: Electrode, Probe, Manual, 50mL refill bottle

    Specifications

    Connector Types BNC/2.5mm plug
    Storage Requirements Ambient temperature
    Filling Solution Type 3M KCl Potassium Chloride
    Reference Type Iodine/Iodide
    Diameter (Metric) Body 5mm
    Junction 3X Junction
    Material Glass

    Up your performance and accuracy with GoldLINE. The unique three-chamber reference system is more stable with a metal-free iodide/iodine reference system compared to our excellent Ag/AgCl reference system found in our PlatiumLINE series. GoldLINE not only offers unbeatable stability and faster response times but higher accuracy and precision. Accelerated performance is attributed to the advanced patented reference technologies and superior glass and reference materials & construction. The GoldLINE is a superior sensor that gives precise results allowing for independence from sample contamination with minimal temperature influences.

    • GoldLINE electrodes have lower temperature sensitivity, reducing the drastic influences temperature variants can have on pH measurement, thus producing more precise results.
    • Exchangeable bridge electrolyte enables one to match the internal electrolyte to their sample for better measurements.
    • Speed and response time is quicker due to stability of reference exchange, system and damping of temperature influences and the reduction of metal or precipitant contamination that can occur with other electrodes.

    Patented junction and three-chamber systems set sensors apart from the rest
    • Reduce and eliminate metal ion contamination and chelation
    • Minimizes damage to sensors elongating the life of the electrodes, saving time and money as well as increased productivity to mitigate faulty measurements

    Smart ID sensor versions
    • A reliable and accurate measurement through automatic electrode recognition between sensor and meter
    • Provide unique identification for each sensor for traceability
    • Can be used on multiple instruments without having to recalibrate
    • Stores the last calibration of the sensor
    • Completes the documentation trail between sensor and meter for the highest GLP practices

    • Electrode includes model, serial number with date, time and measurement values


    PY-P28-2S
    Pharma Grade – pH/ATC, TRIS Impervious, Glass Body, Liquid Filled 3M KCl, 3X Junction, Iodine/Iodide reference

    PY-P29-2S
    Micro – 5mm dia., pH/ATC, TRIS Impervious, Glass Body, Liquid Filled 3M KCl, 3X Junction, Iodine/Iodide-reference

    PY-P30-2S
    Speartip pH/ATC Glass Body, 3x, Hole Junction, Polymer Filled

    Warranty

    1 year

  • Description & Specifications

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit