Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ GOLGA6L9 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA549041
Description
GOLGA6L9 Polyclonal Antibody for Western Blot

Specifications
GOLGA6L9 | |
Polyclonal | |
Unconjugated | |
GOLGA6L9 | |
GOLGA6L20; GOLGA6L9; golgi autoantigen, golgin subfamily a, 6-like 9; golgin A6 family-like 20; golgin A6 family-like 9; golgin subfamily A member 6-like protein 9; putative golgin subfamily A member 6-like protein 20 | |
Rabbit | |
Antigen affinity chromatography, Protein A | |
RUO | |
440295 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Western Blot | |
0.5 mg/mL | |
PBS with 0.09% sodium azide; pH 7.4 | |
A6NEM1 | |
GOLGA6L9 | |
KLH conjugated synthetic peptide between 15-41 amino acids from the N-terminal region of human GOLGA6L9. | |
400 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction