Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Golgi Complex Rabbit anti-Human, Clone: GLG1/2829R, Novus Biologicals™
Rabbit Monoclonal Antibody
Supplier: Novus Biologicals NBP30727810UG
Description
Golgi Complex Monoclonal specifically detects Golgi Complex in Human samples. It is validated for Immunohistochemistry-Paraffin.Specifications
Golgi Complex | |
Monoclonal | |
0.2 mg/ml | |
Immunohistochemistry-Paraffin 1-2 ug/ml | |
Rabbit | |
Protein A or G purified | |
RUO | |
Human | |
Purified |
Immunohistochemistry (Paraffin) | |
GLG1/2829R | |
Unconjugated | |
10 mM PBS with 0.05% BSA | |
SU-DHL-1 large cell lymphoma cells. | |
100 μg | |
Primary | |
Store at 4°C. | |
IgG |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title