Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

GOLM1, Mouse anti-Human, Clone: 6C9, Abnova™

Mouse Monoclonal Antibody

Manufacturer:  Abnova Corporation MAB16354

Catalog No. 89-999-112


Add to cart

Description

Description

Specifications

Specifications

GOLM1
6C9
Immunofluorescence (1:100) Immunohistochemistry (Formalin/PFA-fixed paraffin-embedded sections) (10 ug/mL) Western Blot (1:500-1:2000) The optimal working dilution should be determined by the end user.
golgi membrane protein 1
C9orf155/FLJ22634/FLJ23608/GOLPH2/GP73/PSEC0257/bA379P1.3
Mouse
IgG1
RUO
Antibody
Monoclonal
Human
Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot
Unconjugated
In PBS, pH 7.3 (50% glycerol, 1% BSA, 0.02% sodium azide).
Liquid
GOLM1
Recombinant protein corresponding to full length human GOLM1 from HEK293T cell.
50 μL
Store at -20°C. Aliquot to avoid repeated freezing and thawing.
Primary
51280
SDS
Documents
Product Certifications
Promotions

Promotions

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit