Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

GOLM1 Mouse anti-Human, Clone: OTI3D7, liquid, TrueMAB™ Non-distribution product as customer accommodation.

Mouse Monoclonal Antibody

Manufacturer:  OriGeneSupplier Diversity Partner TA504111

Catalog No. 50-167-4864


Add to cart

Description

Description

GOLPH2, also known as GOLM1, is a Golgi phosphoprotein that has a short cytoplasmic N-terminal domain, a membrane spanning region, and a longer C-terminal domain. It was initially identified as a possible marker for Alzheimer ft.s disease, although later studies have demonstrated that the GOLPH2 gene does not contribute to risk of this disease. GOLPH2 expression has been reported higher in prostate cancer tissues compared to normal prostate tissue, suggesting that GOLPH2 can be used as an additional positive marker for tissue-based diagnosis of prostate cancer. It has been suggested that GOLPH2 expression in hepatocellular carcinomas (HCCs) and serum may also serve as tumor markers for HCCs.
Specifications

Specifications

GOLM1
Monoclonal
0.68 mg/mL
PBS with 1% BSA, 50% glycerol and 0.02% sodium azide
Q8NBJ4
C9orf155, GOLPH2, GP73, HEL46, PSEC0257, bA379P1.3
Mouse
IgG2b
100 μL
-20° C, Avoid Freeze/Thaw Cycles
Primary
51280
Immunohistochemistry (Paraffin), Western Blot
OTI3D7
Unconjugated
GOLM1
Liquid
GOLM1
Full length human recombit protein of human GOLM1 produced in HEK293T cell.
Affinity Chromatography
RUO
Antibody
Monoclonal
Human
Documents
Provide Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit