Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
GOLT1B Rabbit anti-Mouse, Polyclonal, Invitrogen™
Rabbit Polyclonal Antibody
Supplier: Thermo Scientific PA571780
Description
GOT1B may be involved in fusion of ER-derived transport vesicles with the Golgi complex.
Specifications
GOLT1B | |
Polyclonal | |
Unconjugated | |
Golt1b | |
Vesicle transport protein GOT1B; Germ cell tumor 2; Golgi transport 1 homolog B; Putative NF-kappa-B-activating protein 470; hGOT1a; GOLT1B; GCT2; GOT1A | |
Rabbit | |
Protein A | |
RUO | |
66964 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot | |
0.61 mg/mL | |
PBS with 0.09% sodium azide | |
Q9CR60 | |
Golt1b | |
KLH conjugated synthetic peptide between 108-136 amino acids from the C-terminal region of human GOT1B | |
400 μL | |
Primary | |
Mouse | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction