Learn More
MilliporeSigma™ Gomori's Methenamine Silver Plating Kit
Used for detection of argent-affine structures in histological tissue
Supplier:  MilliporeSigma™ 1.00820.0001
Catalog No. M1008200001
This item has been discontinued and is no longer available. Please call customer service for assistance: 1-800-766-7000.
Description
Description
  • Intestifies color of silver deposits by treatment with gold chloride solution
  • Forms silver-gold complex
  • Gold chloride solution prevents non-specific staining
  • Microwave protocol
Specifications
Specifications
Fibre Case
1 Set