Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Gonadotropin Inducible Transcription Repressor 1 Rabbit anti-Human, Polyclonal, Novus Biologicals™

Rabbit Polyclonal Antibody

$106.19 - $352.89

Specifications

Antigen Gonadotropin Inducible Transcription Repressor 1
Dilution Western Blot 1.0 ug/ml
Applications Western Blot
Conjugate Unconjugated
Format Purified
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
NB125959
View Documents
Novus Biologicals
NBP310529100UL
100 μg
Each for $352.89
Add to cart
 
NB125960
View Documents
Novus Biologicals
NBP31052925UL
25 μg
Each for $106.19
Add to cart
 
Description

Description

Gonadotropin Inducible Transcription Repressor 1 Polyclonal antibody specifically detects Gonadotropin Inducible Transcription Repressor 1 in Human samples. It is validated for Western Blot
Specifications

Specifications

Gonadotropin Inducible Transcription Repressor 1
Western Blot
Purified
RUO
PBS buffer, 2% sucrose
92283
Polyclonal
Affinity purified
Western Blot 1.0 ug/ml
Unconjugated
Rabbit
Human
zinc finger protein 213, Zinc finger protein with KRAB and SCAN domains 21, ZKSCAN21CR53Putative transcription factor CR53
The immunogen is a synthetic peptide directed towards the C-terminal region of Human ZN461. Peptide sequence FRLHSHLIQHQRIHTGEKPYECKECGKAFSYHSSFSHHQRIHSGKKPYQC
Primary
Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
SDS
Product Certifications
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit