Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Goosecoid Mouse anti-Human, DyLight 755, Clone: 4C5D5, Novus Biologicals™

Mouse Monoclonal Antibody

Manufacturer:  Novus BiologicalsSupplier Diversity Partner NBP237366IR

Catalog No. NBP237366IR

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.


Specifications

Specifications

Goosecoid
4C5D5
Western Blot, Flow Cytometry, ELISA, Immunohistochemistry
Purified
GSC
Purified recombinant fragment of human GSC (AA: 191-257) expressed in E. Coli.
Protein A or G purified
Store at 4C in the dark.
Monoclonal
Human
ELISA, Flow Cytometry, Immunohistochemistry, Western Blot
DyLight 755
50mM Sodium Borate with 0.05% Sodium Azide
goosecoid homeobox, homeobox protein goosecoid
Mouse
IgG1
0.1 ml
Primary
145258
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit