Learn More
Aviva Systems Biology Corp GOPC Antibody - N-terminal region (ARP56214_P050) | 100uL
Supplier:  Aviva Systems Biology Corp ARP56214_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against GOPC. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the N terminal region of human GOPC. Alternate names: CAL, FIG, GOPC1, PIST, dJ94G16.2ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Human, Mouse, Rabbit, Rat, Zebrafish
Catalog No. 50-014-61762