Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

GORASP1 Mouse anti-Canine, Human, Rat, Clone: OTI5G8, liquid, TrueMAB™ Non-distribution product as customer accommodation.

Mouse Monoclonal Antibody

Manufacturer:  OriGeneSupplier Diversity Partner TA501309

Catalog No. 50-167-2455


Add to cart

Description

Description

Grasp65 is a structural protein of the golgi apparatus. It has been implicated in the stacking of golgi cisternae. Grasp65 is shown to have the properties to bind surfaces together in a mitotically regulated matter.
Specifications

Specifications

GORASP1
Monoclonal
0.6 mg/mL
PBS with 1% BSA, 50% glycerol and 0.02% sodium azide
O35254, Q9BQQ3
GOLPH5, GRASP65, P65
Mouse
IgG1
100 μL
-20° C, Avoid Freeze/Thaw Cycles
Primary
485594, 56082, 64689
Flow Cytometry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot
OTI5G8
Unconjugated
GORASP1
Liquid
Gorasp1
Full length human recombit protein of human GORASP1 produced in HEK293T cell.
Affinity Chromatography
RUO
Antibody
Monoclonal
Canine, Human, Rat
Documents
Provide Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit