Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ GOSR2 Monoclonal Antibody (4HAD6)
Mouse Monoclonal Antibody
Supplier: Invitrogen™ MA125618
Description
Recommended positive controls: HeLa heat shocked lysate.
This gene encodes a trafficking membrane protein which transports proteins among the medial- and trans-Golgi compartments. Due to its chromosomal location and trafficking function, this gene may be involved in familial essential hypertension. Three transcript variants encoding three different isoforms have been found for this gene.
Specifications
GOSR2 | |
Monoclonal | |
1 mg/mL | |
PBS with 50% glycerol, 0.1 mM PMSF and no preservative; pH 7.2 | |
O14653, O35165, O35166 | |
Gosr2 | |
Recombinant rat membrin with a His6 tag. | |
50 μg | |
Primary | |
Human, Mouse, Rat, Canine, Rabbit, Hamster, Pig | |
Antibody | |
IgG1 |
Immunoprecipitation, Western Blot, Immunocytochemistry | |
4HAD6 | |
Unconjugated | |
Gosr2 | |
2310032N09Rik; 27 kDa Golgi SNARE protein; Bos1; C76855; EPM6; Golgi SNAP receptor complex member 2; golgi SNARE; Gosr2; Gs27; I79_025823; LOC100766577; Membrin; RP23-272P17.5; SNARE | |
Mouse | |
Protein G | |
RUO | |
100342992, 100519332, 100766577, 56494, 610436, 64154, 9570 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction