Learn More
Corning™ Petri Dishes
Optimized for automation. Gosselin™ Petri Dishes are manufactured from crystal polystyrene.
Manufacturer:  Corning™ SB9303
 View more versions of this product
Catalog No. 07-201-914
This item has been discontinued and is no longer available. Please call customer service for assistance: 1-800-766-7000.
View substitute products »
This item is not returnable. View return policy
Description
Description
Optimized for automation. Gosselin™ Petri Dishes are manufactured from crystal polystyrene.
  • Crystal polystyrene
  • Optimal flatness
  • Manufactured in class 100
  • Easy opening
Specifications
Specifications
90 mm
Sterile
1
Regular
Round
3
15 mm
Transparent
Polystyrene