Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Gosselin™ Sterile Polypropylene Dippers with Removable Handle Available on GSA/VA Contract for Federal Government customers only.

Quality sampling tools for secure sample retrieval and shipment

Manufacturer:  Gosselin TAT5201

 View more versions of this product

Catalog No. 07-201-973

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
View substitute products »


Specifications

Description

  • Polypropylene blue containers
  • Removable handle
  • Shock resistant, Leakproof
  • No cross contamination risks
  • Traceability information on each bag
  • Packaged individually in tamper evident single zipped bags

Description & Specifications

Specifications

4.22 oz., 125mL
Blue
5.2cm
10.82 in.
Removable handle
Yes
125mL
EC N 1935/2004, ISO 11137
2.04 in.
Quality sampling tools for secure sample retrieval and shipment
27.5cm
Polypropylene
4.22 oz.
Dipper
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit