Learn More
Corning™ Sterile Polypropylene Straight Containers with Red Polyethylene Caps
Quality sampling tools for secure sample retrieval and shipment
Manufacturer:  Corning™ TP35A009
 View more versions of this product
Catalog No. 07-201-936
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
This item is not returnable. View return policy
Description
Description
  • Injection molded polypropylene
  • Leakproof red polyethylene screw cap
  • Assured Sterile 10-3
Specifications
Specifications
Sterile
Clear Polypropylene
2.02 oz.
33mm
2.02 oz., 60mL
Red
Red polyethylene leakproof screw cap
60mL
70mm
Polyethylene