Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ Gossypol-acetic acid

Click to view available options
Quantity:
25mg
100mg
12542-36-8
C32H34O10
578.614
MFCD00058385
NIOHNDKHQHVLKA-UHFFFAOYSA-N
gossypol-acetic acid,gossypol acetate,gossypol acetic acid,acetate gossypol,--gossypol acetic acid,s-gossypol acetic acid,at-101 acetic acid,gossypol-acetic acid complex,s-gossypol acetic acid,at-101 acetic acid
227456
acetic acid;7-(8-formyl-1,6,7-trihydroxy-3-methyl-5-propan-2-ylnaphthalen-2-yl)-2,3,8-trihydroxy-6-methyl-4-propan-2-ylnaphthalene-1-carbaldehyde
CC1=C(C(=C2C(=C1)C(=C(C(=C2C=O)O)O)C(C)C)O)C3=C(C=C4C(=C3O)C(=C(C(=C4C(C)C)O)O)C=O)C.CC(=O)O
12542-36-8
C32H34O10
578.614
MFCD00058385
NIOHNDKHQHVLKA-UHFFFAOYSA-N
gossypol-acetic acid,gossypol acetate,gossypol acetic acid,acetate gossypol,--gossypol acetic acid,s-gossypol acetic acid,at-101 acetic acid,gossypol-acetic acid complex,s-gossypol acetic acid,at-101 acetic acid
227456
acetic acid;7-(8-formyl-1,6,7-trihydroxy-3-methyl-5-propan-2-ylnaphthalen-2-yl)-2,3,8-trihydroxy-6-methyl-4-propan-2-ylnaphthalene-1-carbaldehyde
CC1=C(C(=C2C(=C1)C(=C(C(=C2C=O)O)O)C(C)C)O)C3=C(C=C4C(=C3O)C(=C(C(=C4C(C)C)O)O)C=O)C.CC(=O)O
Color Yellow to Green
Quantity 25mg
Formula Weight 578.61
Physical Form Powder
Chemical Name or Material Gossypol-acetic acid

GHS H Statement
H351-H302
Suspected of causing cancer.
Harmful if swallowed.

GHS P Statement
P281-P264-P301+P312-P308+P313-P405-P501a
Use personal protective equipment as required.
Wash thoroughly after handling.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF exposed or concerned: Get medical advice/attention.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning

RTECSNumber : MD7360000

storageNote1 : -30° to -10°C