Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

(±)-Gossypol-acetic acid, ˜95% (HPLC); ˜95%, MilliporeSigma™ Supelco™

$179.70 - $538.00

Specifications

Percent Purity 95% (HPLC); 95%
Grade Analytical Standard
Linear Formula C30H30O8 · C2H4O2
Physical Form Neat
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
11-101-2846
View Documents
MilliporeSigma Supelco
G4382250MG
250 mg
Each for $179.70
Add to cart
 
11-101-2845
View Documents
MilliporeSigma Supelco
G43821G
1 g
Each for $538.00
Add to cart
 
Description

Description

Featured Industries: Food and Beverages
Specifications

Specifications

95% (HPLC); 95%
Analytical Standard
C30H30O8 · C2H4O2
Neat
Safety and Handling

Safety and Handling

RTECSNumber : MD7360000

SDS
Product Certifications
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit