Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

GOT1L1 Rabbit anti-Human, Mouse, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA548278

Catalog No. PA548278

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.


Specifications

Specifications

GOT1L1
Polyclonal
PBS with 0.09% sodium azide
Q8NHS2, Q7TSV6
Putative aspartate aminotransferase, cytoplasmic 2, 261-, Glutamate oxaloacetate transaminase 1-like protein 1, Transaminase A-like protein 1, GOT1L1
Rabbit
IgG
400 μL
-20° C, Avoid Freeze/Thaw Cycles
Primary
137362, 76615
Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot
Unconjugated
GOT1L1
Liquid
GOT1L1
KLH conjugated synthetic peptide between 346-375 amino acids from the C-terminal region of human GOT1L1.
Antigen affinity chromatography
RUO
Antibody
Polyclonal
Human, Mouse
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit