Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Novus Biologicals™ GOT2 Human Recombinant Protein
Highly purified and high bioactivity. Generating reliable and reproducible results. Applications: Functional, SDS-Page, Bioactivity
$206.00 - $436.00
Specifications
Concentration | 0.5mg/mL |
---|---|
For Use With (Application) | SDS-Page |
Formulation | 20mM Tris-HCl buffer (pH 8.0) containing 10% glycerol, 1mM DTT, 0.15M NaCl |
Gene ID (Entrez) | 2806 |
Molecular Weight | TMW: 47kDa |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
NBP2521462
|
Novus Biologicals™
NBP2521460.02MG |
0.02mg |
Each for $206.00
|
|
NBP25214601
|
Novus Biologicals™
NBP2521460.1MG |
0.1mg |
Each for $436.00
|
|
Description
A bioactive recombinant protein with a N-Terminal His-tag and corresponding to the amino acids 30-430 of Human GOT2 The Recombinant Human GOT2 Protein is derived from E. coli. The Recombinant Human GOT2 Protein has been validated for the following applications: Functional, SDS-Page, Bioactivity.Specifications
0.5mg/mL | |
20mM Tris-HCl buffer (pH 8.0) containing 10% glycerol, 1mM DTT, 0.15M NaCl | |
TMW: 47kDa | |
Store at 4°C short term. Aliquot and store at −20°C long term. Avoid freeze-thaw cycles. | |
GOT2 | |
Unlabeled |
SDS-Page | |
2806 | |
>95%, by SDS-PAGE under reducing conditions and visualized by Colloidal Coomassie™ Blue stain | |
aspartate aminotransferase, mitochondrial, EC 2.6.1, EC 2.6.1.1, FABP-1, FABPpm, Fatty acid-binding protein, FLJ40994, Glutamate oxaloacetate transaminase 2, glutamic-oxaloacetic transaminase 2, mitochondrial (aspartate aminotransferase2), KAT4, KATIV, kynurenine aminotransferase IV, mAspAT, mitAAT, Plasma membrane-associated fatty acid-binding protein, Transaminase A | |
Specific activity is > 60 units/mg, and is defined as the amount of enzyme that convert 1umole of alpha-ketoglutarate to L-Glutamate per minute at pH 8.0 at 25C. | |
Yes |
For research use only and is not approved for use in humans or in clinical diagnosis
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title