Learn More
BioVision IncSupplier Diversity Partner Glucose Oxidase Activity Colorimetric/Fluorometric Assay Kit: Colorimetric and Fluorometric Assay for Detecting GOx Activity in Cells, Tissues, Serum etc. Simple, Sensitive & Accurate. Detection Limit: 0.01 mU. 100 Assays. DFS Item
Manufacturer:  BioVision IncSupplier Diversity Partner K788100
The glucose oxidase enzyme (GOx) (EC 1.1.3.4) is an oxido-reductase that catalyzes the oxidation of glucose to hydrogen peroxide and D-glucono-δ-lactone. In cells, it aids in breaking the sugar down into its metabolites. BioVision’s Glucose Oxidase Assay Kit provides a convenient tool for sensitive detection of the Glucose Oxidase in a variety of samples. The Glucose Oxidase in sample will recognize d-glucose as a specific substrate leading to proportional color development. The activity of GOx can be easily quantified colorimetrically (λ = 570 nm) or fluorometrically (Ex/Em = 535/585 nm). This assay detects GOx activity as low as 0.01mU with our unit definition. BioVision charges an Ice pack and Insulated Container fee of $25.00 for each order.
Catalog No. 50-111-9906