Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

GP 2a, Tocris Bioscience™

Click to view available options
Quantity:
10mg
50mg
This item is not returnable. View return policy
919077-81-9
C24H23Cl2N3O
440.368
SPEFJYZGXZENAF-UHFFFAOYSA-N
gp 2a,n-cyclohexyl-1-2,4-dichlorophenyl-1,4-dihydro-6-methylindeno 1,2-c pyrazole-3-carboxamide,n-cyclohexyl-1-2,4-dichlorophenyl-6-methyl-1,4-dihydroindeno 1,2-c pyrazole-3-carboxamide,1-2,4-dichlorophenyl-6-methyl-n-cyclohexylamine-1,4-dihydroindeno 1,2-c pyrazole-3-carboxamide,1-2,4-dichlorophenyl-n-cyclohexyl-6-methyl-1,4-dihydroindeno 1,2-c pyrazole-3-carboxamide
16094766
N-cyclohexyl-1-(2,4-dichlorophenyl)-6-methyl-4H-indeno[1,2-c]pyrazole-3-carboxamide
CC1=CC2=C(C=C1)C3=C(C2)C(=NN3C4=C(C=C(C=C4)Cl)Cl)C(=O)NC5CCCCC5
This item is not returnable. View return policy
919077-81-9
C24H23Cl2N3O
440.368
SPEFJYZGXZENAF-UHFFFAOYSA-N
gp 2a,n-cyclohexyl-1-2,4-dichlorophenyl-1,4-dihydro-6-methylindeno 1,2-c pyrazole-3-carboxamide,n-cyclohexyl-1-2,4-dichlorophenyl-6-methyl-1,4-dihydroindeno 1,2-c pyrazole-3-carboxamide,1-2,4-dichlorophenyl-6-methyl-n-cyclohexylamine-1,4-dihydroindeno 1,2-c pyrazole-3-carboxamide,1-2,4-dichlorophenyl-n-cyclohexyl-6-methyl-1,4-dihydroindeno 1,2-c pyrazole-3-carboxamide
16094766
N-cyclohexyl-1-(2,4-dichlorophenyl)-6-methyl-4H-indeno[1,2-c]pyrazole-3-carboxamide
CC1=CC2=C(C=C1)C3=C(C2)C(=NN3C4=C(C=C(C=C4)Cl)Cl)C(=O)NC5CCCCC5

Selective CB2 agonist

Selective CB 2 receptor agonist; K i values are 7.6 and 900 nM for CB 2 and CB 1 receptors respectively. Increases P-ERK1/2 expression in HL-60 cells in vitro.
Quantity 50mg
Assay Percent Range >99%
Formula Weight 440.36
Chemical Name or Material GP 2a

Recommended Storage : Store at Room Temperature