Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

gp130/CD130 Rabbit anti-Rat, Biotin, Clone: 108, Novus Biologicals™

Rabbit Monoclonal Antibody

Supplier:  Novus Biologicals NBP290852B

Catalog No. NB078938


Add to Cart

Description

Description

gp130/CD130 Monoclonal specifically detects gp130/CD130 in Rat samples. It is validated for Flow Cytometry, ELISA.
Specifications

Specifications

gp130/CD130
Monoclonal
Biotin
CD130, CD130 antigen, CDw130, DKFZp564F053, gp130, gp130 of the rheumatoid arthritis antigenic peptide-bearing soluble form, IL-6 receptor subunit beta, IL-6R subunit beta, IL-6RB, IL-6R-beta, interleukin 6 signal transducer (gp130, oncostatin M receptor), interleukin receptor beta chain, interleukin-6 receptor subunit beta, Interleukin-6 signal transducer, Membrane glycoprotein 130, membrane glycoprotein gp130, Oncostatin-M receptor subunit alpha
This antibody was obtained from a rabbit immunized with purified, recombinant Rat gp130/CD130 (NP_001008725.2; Met4-Glu618).
0.1 mL
Cancer, Immunology
3572
Store at 4°C in the dark.
IgG
Flow Cytometry, ELISA
108
PBS
Rabbit
Protein A purified
RUO
Primary
Rat
Purified
SDS
Documents

Documents

Product Certifications
Promotions

Promotions

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit