Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

GP2 Mouse anti-Human, Allophycocyanin/Cy7, Clone: GP2/1551, Novus Biologicals™

Mouse Monoclonal Antibody

Manufacturer:  Novus Biologicals NBP254410APCCY7

Catalog No. N254410APC7


Add to cart

Description

Description

GP2 Monoclonal antibody specifically detects GP2 in Human samples. It is validated for Flow Cytometry.
Specifications

Specifications

GP2
GP2/1551
Flow Cytometry
Conjugated, Purified
GP2
Recombinant human GP2 protein fragment (aa35-179) (exact sequence is proprietary)
Protein A or G purified
Store at 4°C in the dark. Do not freeze.
Monoclonal
Human
Flow Cytometry
APC-Cyanine7
PBS with 0.05% Sodium Azide
DKFZp779K0533, glycoprotein 2 (zymogen granule membrane), pancreatic secretory granule membrane major glycoprotein GP2, pancreatic zymogen granule membrane associated protein GP2, Pancreatic zymogen granule membrane protein GP-2, ZAP75
Mouse
IgG2b Lambda
0.1 mL
Primary
2813.0
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit