Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
GP2 Mouse anti-Human, Clone: GP2/1551, Novus Biologicals™
Mouse Monoclonal Antibody
Specifications
Antigen | GP2 |
---|---|
Clone | GP2/1551 |
Concentration | 0.2 mg/mL |
Dilution | Flow Cytometry 0.5 - 1 ug/million cells, ELISA 1:100 - 1:2000, Immunocytochemistry/Immunofluorescence 0.5 - 1 ug/ml, SDS-Page |
Applications | Flow Cytometry, ELISA, Immunocytochemistry, Immunofluorescence, SDS-Page |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
NB253184200
|
Novus Biologicals
NBP253184200UG |
200 μg | Item Discontinued This item has been discontinued by the supplier. Please Sign In to view product availability in your area. | N/A | |||||
Description
GP2 Monoclonal antibody specifically detects GP2 in Human samples. It is validated for Flow Cytometry, ELISA, Immunocytochemistry/Immunofluorescence, SDS-Page.Specifications
GP2 | |
0.2 mg/mL | |
Flow Cytometry, ELISA, Immunocytochemistry, Immunofluorescence, SDS-Page | |
Unconjugated | |
Mouse | |
Human | |
DKFZp779K0533, glycoprotein 2 (zymogen granule membrane), pancreatic secretory granule membrane major glycoprotein GP2, pancreatic zymogen granule membrane associated protein GP2, Pancreatic zymogen granule membrane protein GP-2, ZAP75 | |
GP2 | |
IgG2b λ | |
Protein A or G purified | |
59 kDa |
GP2/1551 | |
Flow Cytometry 0.5 - 1 ug/million cells, ELISA 1:100 - 1:2000, Immunocytochemistry/Immunofluorescence 0.5 - 1 ug/ml, SDS-Page | |
Monoclonal | |
Purified | |
RUO | |
10mM PBS with 0.05% BSA with 0.05% Sodium Azide | |
2813 | |
Recombinant human GP2 protein fragment (aa35-179) (exact sequence is proprietary) | |
Primary | |
Store at 4C. |
For Research Use Only
Spot an opportunity for improvement?
Provide Content Correction
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title