Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

GP2 Rabbit anti-Human, Clone: GP2/2569R, Novus Biologicals™

Rabbit Monoclonal Antibody

Specifications

Antigen GP2
Clone GP2/2569R
Concentration 0.2 mg/ml
Dilution Western Blot 1-2 ug/ml, ELISA 2-4 ug/ml, Immunohistochemistry-Paraffin 1-2 ug/ml
Applications Western Blot, ELISA, Immunohistochemistry (Paraffin)
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
NB122150
View Documents
Novus Biologicals
NBP30733820UG
20 μg N/A
NB122149
View Documents
Novus Biologicals
NBP30733810UG
100 μg N/A
Description

Description

GP2 Monoclonal antibody specifically detects GP2 in Human samples. It is validated for Western Blot, ELISA, Immunohistochemistry (Paraffin)
Specifications

Specifications

GP2
0.2 mg/ml
Western Blot, ELISA, Immunohistochemistry (Paraffin)
Purified
RUO
10 mM PBS with 0.05% BSA
2813
IgG
Primary
Store at 4°C.
GP2/2569R
Western Blot 1-2 ug/ml, ELISA 2-4 ug/ml, Immunohistochemistry-Paraffin 1-2 ug/ml
Unconjugated
Rabbit
Human
DKFZp779K0533, glycoprotein 2 (zymogen granule membrane), pancreatic secretory granule membrane major glycoprotein GP2, pancreatic zymogen granule membrane associated protein GP2, Pancreatic zymogen granule membrane protein GP-2, ZAP75
Recombinant human GP2 protein fragment (around aa35-179) (exact sequence is proprietary)
Monoclonal
Protein A or G purified
SDS
Product Certifications
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit