Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
GP2 Rabbit anti-Human, Unconjugated, Clone: GP2/2569R, Abnova
Rabbit Monoclonal Antibody.
Manufacturer: Abnova Corporation RAB00420
Description
GP2 Monoclonal antibody specifically detects GP2 in Human samples. It is validated for ELISA, IHC-P.Specifications
GP2 | |
GP2/2569R | |
glycoprotein 2 (zymogen granule membrane) | |
Liquid | |
Rabbit | |
IgG | |
Store at -20 to -80°C. Aliquot to avoid repeated freezing and thawing. | |
Monoclonal | |
Human |
ELISA, Immunohistochemistry (Paraffin) | |
Unconjugated | |
GP2 | |
DKFZp779K0533, ZAP75 | |
Original antibody is raised against recombinant protein corresponding to amino acid 35-179 of human GP2 | |
100 μg | |
Primary | |
2813 |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title