Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

GPAA1 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech

Rabbit Polyclonal Antibody

$395.30

Specifications

Antigen GPAA1
Concentration 0.13 mg/mL
Applications Immunohistochemistry (Paraffin), Western Blot
Conjugate Unconjugated
Format Liquid
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
50-172-6026
Encompass_Preferred
View Documents
Proteintech Group IncSupplier Diversity Partner
101041AP150UL
150 μL
Each for $395.30
Add to cart
 
Description

Description

This antibody is specific to GPAA1.

Posttranslational glycosylphosphatidylinositol (GPI) anchor attachment serves as a general mechanism for linking proteins to the cell surface membrane. The protein encoded by this gene presumably functions in GPI anchoring at the GPI transfer step. The mRNA transcript is ubiquitously expressed in both fetal and adult tissues. The anchor attachment protein 1 contains an N-terminal signal sequence, 1 cAMP- and cGMP-dependent protein kinase phosphorylation site, 1 leucine zipper pattern, 2 potential N-glycosylation sites, and 8 putative transmembrane domains.
Specifications

Specifications

GPAA1
Immunohistochemistry (Paraffin), Western Blot
Liquid
RUO
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3
Polyclonal
-20°C
0.13 mg/mL
Unconjugated
Rabbit
Human, Mouse, Rat
IgG
Antigen Affinity Chromatography
Antibody
SDS
Product Certifications
Provide Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit