Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
GPAT2 Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody has been used in 2 publications
$382.00 - $610.00
Specifications
Antigen | GPAT2 |
---|---|
Dilution | Immunohistochemistry, Immunocytochemistry/Immunofluorescence 1-4 ug/ml, Immunohistochemistry-Paraffin 1:50 - 1:200 |
Applications | Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) |
Classification | Polyclonal |
Conjugate | Unconjugated |
Description
GPAT2 Polyclonal specifically detects GPAT2 in Human samples. It is validated for Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.Specifications
GPAT2 | |
Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
Unconjugated | |
RUO | |
PBS (pH 7.2) and 40% Glycerol with 0.02% Sodium Azide | |
150763 | |
This antibody was developed against Recombinant Protein corresponding to amino acids:HQKGVFLSQLLGEFSWLTEEILLRGFDVGFSGQLRSLLQHSLSLLRAHVALLRIRQGDLLVVPQPGPGLTHLA | |
Primary | |
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
Immunohistochemistry, Immunocytochemistry/Immunofluorescence 1-4 ug/ml, Immunohistochemistry-Paraffin 1:50 - 1:200 | |
Polyclonal | |
Rabbit | |
Human | |
ALPAAB, BSCL, BSCL1, cancer/testis antigen 123, CT123, EC 2.3.1.15, glycerol-3-phosphate acyltransferase 2, mitochondrial, GPAT-2, xGPAT1 | |
GPAT2 | |
IgG | |
Affinity Purified | |
Specificity of antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title