Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ GPATCH2L Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA571419
Description
GPATCH2L Polyclonal Antibody for Western Blot
GPATCH2L is a protein coding gene.
Specifications
GPATCH2L | |
Polyclonal | |
Unconjugated | |
GPATCH2L | |
1700020O03Rik; 4732487G21Rik; A630090I05Rik; C10H14orf118; C14orf118; C8H14orf118; G patch domain containing 2 like; G patch domain containing 2-like; G patch domain-containing protein 2-like; G-patch domain containing 2 like; GPATCH2L; Kiaa1152; RGD1308907 | |
Rabbit | |
Affinity chromatography | |
RUO | |
55668 | |
-20°C, Avoid Freeze/Thaw Cycles | |
Liquid |
Western Blot | |
0.5 mg/mL | |
PBS with 2% sucrose and 0.09% sodium azide | |
Q9NWQ4 | |
GPATCH2L | |
synthetic peptide directed towards the middle region of human GPATCH2L. | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction