Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

GPBP Rabbit anti-Human, Polyclonal, Novus Biologicals™

Rabbit Polyclonal Antibody

Manufacturer:  Novus Biologicals NBP18001220UL

 View more versions of this product

Catalog No. NBP1800220


Add to cart

Description

Description

GPBP Polyclonal antibody specifically detects Antigen in Human samples. It is validated for Immunohistochemistry, Immunohistochemistry (Paraffin), Western Blotting.
Specifications

Specifications

GPBP
Unconjugated
PBS & 2% Sucrose. with No Preservative
Affinity Purified
GPBP1
Synthetic peptide directed towards the N terminal of human DKFZP761C169. Peptide sequence RKEKNGWRTHGRNGTENINHRGGYHGGSSRSRSSIFHAGKSQGLHENNIP.
Immunogen affinity purified
RUO
Primary
65056
Immunohistochemistry, Immunohistochemistry (Paraffin), Western Blot
Western Blot 1:1000, Immunohistochemistry 1:10-1:500, Immunohistochemistry-Paraffin 1:10-1:500
NP_075064
GC-rich promoter binding protein 1, MGC126339, SSH6, vascular wall-linked protein
Rabbit
IgG
20ul
Store at -20C. Avoid freeze-thaw cycles.
Polyclonal
Human
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit