Learn More
GPBP1 Rabbit anti-Human, Polyclonal, Abnova™
Rabbit Polyclonal Antibody
Manufacturer:  Abnova Corporation PAB30108
Catalog No. 89-033-396
Add to cart
Description
Description
Rabbit polyclonal antibody raised against synthetic peptide of human GPBP1.
  • High specificity
  • High reactivity
Specifications
Specifications
GPBP1
Unconjugated
Immunohistochemistry (Formalin/PFA-fixed paraffin-embedded sections) (4-8 ug/mL) Western Blot (0.2-1 ug/mL) The optimal working dilution should be determined by the end user.
NM_022913
DKFZp761C169|GPBP|MGC126339|VASCULIN
Rabbit
Affinity purification
RUO
Primary
65056
Immunohistochemistry (PFA fixed), Western Blot
Rabbit polyclonal antibody raised against synthetic peptide of human GPBP1.
In PBS (2% sucrose, 0.09% sodium azide).
Liquid
GPBP1
A synthetic peptide corresponding to N-terminus of human GPBP1.
100 uL
Store at 4°C. For long term storage store at -20°C.
Aliquot to avoid repeated freezing and thawing.
Polyclonal
Human
Documents
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Cancel Submit
For Research Use Only (RUO)