Learn More
Showa Denko America Inc GPC HK-401 DFS Item
Manufacturer:  Showa Denko America Inc F6025010
Newly developed styrene divinylbenzene copolymer with mono-dispersed particle size packing material, analysis time reduced to about one sixth of conventional column's analysis time, low column pressure even under high flow rate, can be used with but does not require a UHPLC system, the amount of solvent used is reduced to about a sixth, HK-HFIP404L is filled with HFIP, fulfills USP L21 requirements, MW range (polystyrene) = 100 − 1,500 Daltons, exclusion limit = 2,000 Daltons, dimensions 4.6 mm ID x 150 mm L, particle size = 3 μm, pore size = 50 Å
Catalog No. 50-161-3297