Learn More
Showa Denko America Inc GPC KF-805 DFS Item
Manufacturer:  Showa Denko America Inc F6028050
Standard THF organic solvent SEC (GPC) column, supports a wide range of applications from low to high molecular weight compounds, fulfills USP L21 requirements, KF-805 MW range (polystyrene) = 50,000 − 2,000,000 Daltons, exclusion limit = 4,000,000 Daltons, dimensions 8.0 mm ID x 300 mm L, particle size = 10 μm, pore size = 5,000 Å
Catalog No. 50-161-3293