Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ GCP6 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA550968
Description
GCP6 / TUBGCP6 Polyclonal Antibody detects endogenous levels of GCP6 / TUBGCP6 protein.
Tubulin Gamma Complex Associated Protein 6 (GCP6) is part of a large multisubunit complex required for microtubule nucleation at the centrosome.
Specifications
GCP6 | |
Polyclonal | |
Unconjugated | |
TUBGCP6 | |
AI843275; D430023H11; Gamma-tubulin complex component 6; GCP6; GCP-6; KIAA1669; MCCRP; MCCRP1; MCPHCR; mKIAA1669; TUBGCP6; tubulin gamma complex associated protein 6; tubulin, gamma complex associated protein 6 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
85378 | |
-20°C, Avoid Freeze/Thaw Cycles | |
Liquid |
ELISA, Immunohistochemistry, Western Blot | |
1 mg/mL | |
PBS with 0.5% BSA, 50% glycerol and 0.02% sodium azide; pH 7.4 | |
Q96RT7 | |
TUBGCP6 | |
Synthesized peptide derived from the Internal region of human GCP6 (aa 741-790). | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction