Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ GPHA2 Monoclonal Antibody (06)
Mouse Monoclonal Antibody
Supplier: Invitrogen™ MA529316
Description
This product is preservative free. It is recommended to add sodium azide to avoid contamination (final concentration 0.05%-0.1%). This antibody has specificity for Human GPHA2/glycoprotein hormone alpha 2.
GPHA2 is a cystine knot-forming polypeptide and a subunit of the dimeric glycoprotein hormone family.
Specifications
GPHA2 | |
Monoclonal | |
1 mg/mL | |
PBS with no preservative | |
Q96T91 | |
Gpha2 | |
Recombinant Human GPHA2/glycoprotein hormone alpha 2 Protein (Met1-Tyr129). | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG1 |
ELISA | |
06 | |
Unconjugated | |
Gpha2 | |
A2; cysteine knot protein; glycoprotein alpha 2; glycoprotein hormone alpha 2; glycoprotein hormone alpha-2; Gpa2; Gpha2; Putative secreted protein Zsig51; thyrostimulin subunit alpha; ZSIG51 | |
Mouse | |
Protein A | |
RUO | |
170589 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction