Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

GPHN Mouse anti-Human, Clone: OTI3C6, liquid, TrueMAB™ Non-distribution product as customer accommodation.

Mouse Monoclonal Antibody

Manufacturer:  OriGeneSupplier Diversity Partner TA502247

Catalog No. 50-167-3321


Add to cart

Description

Description

Gephyrin is a neuronal assembly protein that anchors inhibitory neurotransmitter receptors to the postsynaptic cytoskeleton via high affinity binding to a receptor subunit domain and tubulin dimers. In nonneuronal tissues, the encoded protein is also required for molybdenum cofactor biosynthesis. Mutations in this gene may be associated with the neurological condition hyperplexia and also lead to molybdenum cofactor deficiency. Numerous alternatively spliced transcript variants encoding different isoforms have been described; however, the full-length nature of all transcript variants is not currently known.
Specifications

Specifications

GPHN
Monoclonal
0.83 mg/mL
PBS with 1% BSA, 50% glycerol and 0.02% sodium azide
Q9NQX3
GEPH, GPH, GPHRYN, HKPX1, MOCODC
Mouse
IgG2a
100 μL
-20° C, Avoid Freeze/Thaw Cycles
Primary
10243
Flow Cytometry, Immunohistochemistry (Paraffin), Western Blot
OTI3C6
Unconjugated
GPHN
Liquid
GPHN
Full length human recombit protein of human GPHN produced in HEK293T cell.
Affinity Chromatography
RUO
Antibody
Monoclonal
Human
Documents
Provide Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit