Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abnova™ GPI Recombinant Protein

$463.00 - $713.00

Specifications

Cross Reactivity Human
Format Solution
Formulation 50mM Tris HCl, 10mM reduced Glutathione, pH 8 in the Elution Buffer
For Use With (Application) Antibody Production, Array, Enzyme-linked Immunoabsorbent Assay, Western Blot (Recombinant protein)
Molecular Weight 86.9
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
89-010-142
View Documents
Abnova™
H00002821P0110
10 ug
Each for $463.00
Add to cart
 
89-010-143
View Documents
Abnova™
H00002821P0125
25 ug
Each for $713.00
Add to cart
 
Description

Description

ELISA, Western Blotting (Recombinant Protein), Antibody Production, Protein Array

Specifications

Specifications

Human
50mM Tris HCl, 10mM reduced Glutathione, pH 8 in the Elution Buffer
86.9
8
Glutathione Sepharose 4 Fast Flow
Yes
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
Solution
Antibody Production, Array, Enzyme-linked Immunoabsorbent Assay, Western Blot (Recombinant protein)
GPI (Human) Recombinant Protein (P01)
In vitro wheat germ expression system
12.5% SDS-PAGE Stained with Coomassie Blue.
Wheat Germ (in vitro)
SDS
Product Certifications
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit