Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

GPIP137 Rabbit anti-Human, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA550353

Catalog No. PA550353


Add to cart

Description

Description

The antibody detects endogenous levels of total GPIP137 / CAPRIN1 protein.

GPIP137 may regulate the transport and translation of mRNAs of proteins involved in synaptic plasticity in neurons and cell proliferation and migration in multiple cell types. It is expressed in the cytoplasm.
Specifications

Specifications

GPIP137
Polyclonal
Unconjugated
CAPRIN1
Liquid
CAPRIN1
Fusion protein corresponding to residues near the C terminal of human cell cycle associated protein 1
Antigen affinity chromatography
RUO
Antibody
Polyclonal
Human
Western Blot
0.8 mg/mL
PBS with 50% glycerol and 0.05% sodium azide; pH 7.3
Q14444
M11S1, GPIAP1, RNG105, GPIP137, p137GPI
Rabbit
IgG
100 μL
-20°C
Primary
4076
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit