Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

GPM6A Rabbit anti-Human, Mouse, Rat, Polyclonal, Novus Biologicals™

Rabbit Polyclonal Antibody

$138.37 - $320.71

Specifications

Antigen GPM6A
Applications Immunohistochemistry, Western Blot
Conjugate Unconjugated
Format Purified
Host Species Rabbit
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
NB093197
View Documents
Novus Biologicals
NBP303901100UL
100 μg Each for $320.71
Add to cart
 
NB093198
View Documents
Novus Biologicals
NBP30390120UL
20 μg Each for $138.37
Add to cart
 
Description

Description

GPM6A Polyclonal antibody specifically detects GPM6A in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry
Specifications

Specifications

GPM6A
Unconjugated
Rabbit
Human, Mouse, Rat
glycoprotein M6A, M6a, M6AGPM6, neuronal membrane glycoprotein M6-a
A synthetic peptide corresponding to a sequence within amino acids 1-100 of human GPM6A (NP_005268.1).
Polyclonal
Affinity purified
Immunohistochemistry, Western Blot
Purified
RUO
PBS with 50% glycerol, pH7.3.
2823
IgG
Primary
Store at -20°C. Avoid freeze-thaw cycles.
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit