Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
GPN2 Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
$206.00 - $499.50
Specifications
Antigen | GPN2 |
---|---|
Applications | Western Blot |
Classification | Polyclonal |
Conjugate | Unconjugated |
Host Species | Rabbit |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
NBP15535920
![]() |
Novus Biologicals
NBP15535920UL |
20 μL |
Each for $206.00
|
|
|||||
NBP155359
![]() |
Novus Biologicals
NBP155359 |
100 μL |
Each for $499.50
|
|
|||||
Description
GPN2 Polyclonal specifically detects GPN2 in Human samples. It is validated for Western Blot.Specifications
GPN2 | |
Polyclonal | |
Rabbit | |
Q9H9Y4 | |
54707 | |
Synthetic peptides corresponding to GPN2 (GPN-loop GTPase 2) The peptide sequence was selected from the middle region of GPN2. Peptide sequence VLQAVDKANGYCFRAQEQRSLEAMMSAAMGADFHFSSTLGIQEKYLAPSN. | |
Primary |
Western Blot | |
Unconjugated | |
RUO | |
ATP binding domain 1 family, member B, ATPBD1B, ATP-binding domain 1 family member B, FLJ10349, GPN-loop GTPase 2 | |
GPN2 | |
IgG | |
34 kDa |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title