Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
GPP 78 hydrochloride, Tocris Bioscience™
Click to view available options
Quantity:
10 mg
50 mg
Description
Inhibitor of nicotinamide phosphoribosyltransferase (NAMPT); potently depletes NAD levels and exhibits cytotoxicity in vitro (IC50 values are 3.0 and 3.8 nM respectively). Also shown to induce autophagy in neuroblastoma cells.
Specifications
Quantity | 10 mg |
Formula Weight | 476.01 |
Percent Purity | >98% |
Chemical Name or Material | GPP 78 hydrochloride |
Safety and Handling
Recommended Storage | Store at -20°C |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction