Learn More
GPR101 Mouse anti-Human, Alexa Fluor 700, Clone: 909603, R&D Systems™
Mouse MonoclonalAntibody
Manufacturer:  R&D Systems FAB7495N100UG
Catalog No. FAB7495N100
Add to cart
This item is not returnable. View return policy
Description
Description
GPR101 Monoclonal antibody specifically detects GPR101 in Human samples. It is validated for Flow Cytometry.
Specifications
Specifications
GPR101
Monoclonal
Alexa Fluor 700
Supplied 0.2 mg/mL in a saline solution containing BSA and Sodium Azide.
G protein-coupled receptor 101, GPCR6, probable G-protein coupled receptor 101
NS0 mouse myeloma cell line transfected with human GPR101 Met1-Pro508, Accession # NP_473362
100 ug
Store the unopened product at 2 - 8 degreesC. Do not use past expiration date.
Monoclonal
Detects human GPR101 in Western blots.
Flow Cytometry
909603
Flow Cytometry 0.25-1 ug/10^6 cells
NP_473362
Mouse
IgG
RUO
Primary
83550
Human
SDS
Documents
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Cancel Submit
For Research Use Only (RUO)