Learn More
GPR115 Mouse anti-Human, Alexa Fluor 405, Clone: 527018, R&D Systems™
Mouse Monoclonal Antibody
Manufacturer:  R&D Systems FAB54371V100UG
Catalog No. FAB54371V10
Add to cart
This item is not returnable. View return policy
Description
Description
GPR115 Monoclonal specifically detects GPR115 in Human samples. It is validated for Flow Cytometry.
Specifications
Specifications
GPR115
527018
Flow Cytometry 0.25-1 ug/106 cells
Q8IZF3
FLJ38076, G protein-coupled receptor 115, G-protein coupled receptor PGR18, PGR18, PGR18G-protein coupled receptor 115, probable G-protein coupled receptor 115, seven transmembrane helix receptor
Chinese hamster ovary cell line CHO-derived recombinant human GPR115, Ser22-Ala347, Accession # Q8IZF3
Protein A or G purified from hybridoma culture supernatant
RUO
Primary
221393
Flow Cytometry
Alexa Fluor 405
Supplied 0.2 mg/mL in a saline solution containing BSA and Sodium Azide.
Purified
Mouse
IgG1
100 μg
Protect from light. Do not freeze. 12 months from date of receipt, 2 to 8 °C as supplied.
Monoclonal
Human